siRNA Details
| Gene Symbol: C8orf87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242668.1 | NP_001229597.1 | 2, 3 | 191 | ||
| GenBank mRNA | BC089453.1 | - | 2, 3 | 191 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C8orf87| Gene Symbol: C8orf87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242668.1 | NP_001229597.1 | 2, 3 | 191 | ||
| GenBank mRNA | BC089453.1 | - | 2, 3 | 191 | ||