siRNA Details
| Gene Symbol: CNTNAP3P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_111893.2 | 18 | 2926 | |||
| Gene Symbol: LOC101930090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017015388.1 | XP_016870877.1 | 9 | 1411 | ||

CNTNAP3P2
Genomic Map