siRNA Details
| Gene Symbol: GLIDR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015363.2 | 2 | 2061 | |||
| NR_126044.1 | 2 | 2043 | ||||
| NR_126045.1 | 3 | 1319 | ||||
| GenBank mRNA | AK092305.1 | - | 2 | 1960 | ||
| BC000869.1 | - | 3 | 1172 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GLIDR| Gene Symbol: GLIDR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015363.2 | 2 | 2061 | |||
| NR_126044.1 | 2 | 2043 | ||||
| NR_126045.1 | 3 | 1319 | ||||
| GenBank mRNA | AK092305.1 | - | 2 | 1960 | ||
| BC000869.1 | - | 3 | 1172 | |||