siRNA Details
| Gene Symbol: SPATA31D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207416.2 | NP_997299.2 | 4 | 4017 | ||
| GenBank mRNA | AK126070.1 | - | 1 | 2721 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPATA31D3| Gene Symbol: SPATA31D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207416.2 | NP_997299.2 | 4 | 4017 | ||
| GenBank mRNA | AK126070.1 | - | 1 | 2721 | ||