siRNA Details
Gene Symbol: C9orf172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080482.2 | NP_001073951.2 | 1 | 2915 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C9orf172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080482.2 | NP_001073951.2 | 1 | 2915 |