siRNA Details
| Gene Symbol: UBE2NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012989.2 | NP_001013007.1 | 1 | 367 | ||
| NR_121210.1 | 1 | 367 | ||||
| GenBank mRNA | GU727624.1 | - | 1 | 367 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBE2NL| Gene Symbol: UBE2NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012989.2 | NP_001013007.1 | 1 | 367 | ||
| NR_121210.1 | 1 | 367 | ||||
| GenBank mRNA | GU727624.1 | - | 1 | 367 | ||