siRNA Details
| Gene Symbol: TRIM77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146162.1 | NP_001139634.1 | 5 | 840 | ||
| NM_001271942.1 | NP_001258871.1 | 4 | 609 | |||
| GenBank mRNA | JF968445.1 | - | 4 | 387 | ||
| JF968446.1 | - | 4 | 258 | |||

TRIM77
Genomic Map