siRNA Details
| Gene Symbol: LOC729159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282301.1 | NP_001269230.1 | 1 | 290 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC729159| Gene Symbol: LOC729159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282301.1 | NP_001269230.1 | 1 | 290 | ||