siRNA Details
| Gene Symbol: LOC107983991 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017027570.1 | XP_016883059.1 | 3 | 442 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107983991| Gene Symbol: LOC107983991 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017027570.1 | XP_016883059.1 | 3 | 442 | ||