siRNA Details
| Gene Symbol: ZNF805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001023563.3 | NP_001018857.2 | 4 | 1785 | ||
| NM_001145078.1 | NP_001138550.1 | 3 | 1658 | |||
| GenBank mRNA | AK297977.1 | - | 3 | 1677 | ||
| BC140786.1 | - | 1 | 924 | 9021654 | ||
| CR936617.1 | - | 1 | 963 | |||

ZNF805
Genomic Map