siRNA Details
| Gene Symbol: FRRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013660.2 | NP_001013682.2 | 15, 16 | 2189 | ||
| GenBank mRNA | AK131302.1 | - | 15, 16 | 2189 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FRRS1| Gene Symbol: FRRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013660.2 | NP_001013682.2 | 15, 16 | 2189 | ||
| GenBank mRNA | AK131302.1 | - | 15, 16 | 2189 | ||