siRNA Details
| Gene Symbol: PTRHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013663.1 | NP_001013685.1 | 1 | 215 | ||
| GenBank mRNA | BC073803.1 | - | 1 | 215 | 4896064 | |
| Gene Symbol: CENPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199803.2 | NP_001186732.1 | 1 | 50 | ||
| NM_001322101.1 | NP_001309030.1 | 1 | 50 | |||
| NM_024322.3 | NP_077298.1 | 1 | 50 | |||
| NR_136182.1 | 1 | 50 | ||||
| NR_136183.1 | 1 | 50 | ||||
| NR_136184.1 | 1 | 50 | ||||
| GenBank mRNA | CF993857.1 | - | 1 | 77 | ||

PTRHD1
Genomic Map