siRNA Details
| Gene Symbol: VENTXP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002311.1 | 1 | 630 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

VENTXP7| Gene Symbol: VENTXP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002311.1 | 1 | 630 | |||