siRNA Details
| Gene Symbol: FAM160A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109977.1 | NP_001103447.1 | 6 | 1407 | ||
| XM_005263199.1 | XP_005263256.1 | 5 | 1282 | |||
| XM_011532220.1 | XP_011530522.1 | 4 | 1160 | |||
| XM_011532221.2 | XP_011530523.1 | 6 | 1480 | |||
| XM_011532222.2 | XP_011530524.1 | 5 | 1402 | |||
| XM_011532223.2 | XP_011530525.1 | 4 | 1277 | |||
| XM_011532224.2 | XP_011530526.1 | 4 | 976 | |||
| GenBank mRNA | BC014937.1 | - | 4 | 991 | ||

FAM160A1
Genomic Map