siRNA Details
| Gene Symbol: FAM160A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109977.1 | NP_001103447.1 | 9 | 1760 | ||
| XM_005263199.1 | XP_005263256.1 | 8 | 1635 | |||
| XM_011532220.1 | XP_011530522.1 | 7 | 1513 | |||
| XM_011532221.2 | XP_011530523.1 | 9 | 1833 | |||
| XM_011532222.2 | XP_011530524.1 | 8 | 1755 | |||
| XM_011532223.2 | XP_011530525.1 | 7 | 1630 | |||
| XM_011532224.2 | XP_011530526.1 | 7 | 1329 | |||
| GenBank mRNA | BC014937.1 | - | 7 | 1344 | ||

FAM160A1
Genomic Map