siRNA Details
Gene Symbol: RP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK098176.1 | - | 13 | 1693 | ||
AK302348.1 | - | 11 | 1194 | |||
AK304443.1 | - | 13 | 1355 | |||
AK316511.1 | - | 13 | 1789 |
Gene Symbol: LOC107984125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017014157.1 | XP_016869646.1 | 6 | 902 | ||
XM_017014158.1 | XP_016869647.1 | 1 | 183 |
