siRNA Details
| Gene Symbol: FAM221B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012446.3 | NP_001012448.2 | 2 | 556 | ||
| NR_052026.1 | 2 | 556 | ||||
| XM_006716768.3 | XP_006716831.1 | 3 | 1103 | |||
| GenBank mRNA | BC144503.1 | - | 3 | 534 | ||
| BC146808.1 | - | 2 | 541 | 8860353 | ||
| BX648702.1 | - | 2 | 556 | |||

FAM221B
Genomic Map