siRNA Details
| Gene Symbol: CTSL3P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027917.1 | 3 | 252 | |||
| GenBank mRNA | AJ851862.1 | - | 3 | 252 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CTSL3P| Gene Symbol: CTSL3P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027917.1 | 3 | 252 | |||
| GenBank mRNA | AJ851862.1 | - | 3 | 252 | ||