siRNA Details
Gene Symbol: ZNF254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278661.1 | NP_001265590.1 | 4 | 504 | ||
NM_001278662.1 | NP_001265591.1 | 4 | 484 | |||
NM_001278664.1 | NP_001265593.1 | 2 | 242 | |||
NM_001278665.1 | NP_001265594.1 | 3 | 369 | |||
NM_001278677.1 | NP_001265606.1 | 5 | 576 | |||
NM_203282.3 | NP_975011.3 | 3 | 369 | |||
XM_011528443.1 | XP_011526745.1 | 5 | 637 | |||
XM_011528444.1 | XP_011526746.1 | 6 | 657 | |||
XM_017027514.1 | XP_016883003.1 | 5 | 567 | |||
XM_017027515.1 | XP_016883004.1 | 7 | 994 | |||
XM_017027516.1 | XP_016883005.1 | 3 | 3041 | |||
XM_017027517.1 | XP_016883006.1 | 3 | 797 | |||
XM_017027518.1 | XP_016883007.1 | 3 | 366 | |||
XM_017027519.1 | XP_016883008.1 | 4 | 645 | |||
GenBank mRNA | AF054180.1 | - | 3 | 355 | ||
AK291286.1 | - | 3 | 336 | |||
BC043147.1 | - | 3 | 341 | 5296940 | ||
BC044906.1 | - | 1 | 2425 | |||
BC056243.1 | - | 3 | 345 | |||
BC139749.1 | - | 3 | 300 | 40115903 | ||
BX281205.1 | - | 4 | 372 | |||
DA101675.1 | - | 5 | 494 | |||
DA195951.1 | - | 2 | 100 | |||
DA684942.1 | - | 2 | 225 | |||
DB286139.1 | - | 4 | 468 |
