siRNA Details
| Gene Symbol: MEX3D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174118.1 | NP_001167589.1 | 1, 2 | 594 | ||
| NM_203304.3 | NP_976049.3 | 1, 2 | 594 | |||
| GenBank mRNA | AB107353.1 | - | 1, 2 | 187 | ||
| AF458084.1 | - | 1, 2 | 66 | |||
| AY950680.1 | - | 1, 2 | 594 | |||
| BC113384.1 | - | 1, 2 | 65 | |||
| BC113741.1 | - | 1, 2 | 65 | |||
| DQ225272.1 | - | 2, 3 | 264 | |||
| DQ225273.1 | - | 2, 3 | 225 | |||

MEX3D
Genomic Map