siRNA Details
| Gene Symbol: MEX3D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174118.1 | NP_001167589.1 | 2 | 1371 | ||
| NM_203304.3 | NP_976049.3 | 2 | 1371 | |||
| XM_017026811.1 | XP_016882300.1 | 2 | 1171 | |||
| GenBank mRNA | AB107353.1 | - | 2 | 964 | ||
| AF458084.1 | - | 2 | 843 | |||
| AY950680.1 | - | 2 | 1371 | |||
| BC113384.1 | - | 2 | 842 | |||
| BC113741.1 | - | 2 | 842 | |||

MEX3D
Genomic Map