siRNA Details
| Gene Symbol: FAM102A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035254.2 | NP_001030331.1 | 6 | 869 | ||
| NM_203305.2 | NP_976050.1 | 3 | 351 | |||
| GenBank mRNA | BC047949.1 | - | 3 | 351 | ||
| BC137087.1 | - | 3 | 225 | 9021084 | ||
| BC137088.1 | - | 3 | 225 | 9021085 | ||
| CN482844.1 | - | 6 | 552 | |||

FAM102A
Genomic Map