siRNA Details
| Gene Symbol: FAM102A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035254.2 | NP_001030331.1 | 7 | 1028 | ||
| NM_203305.2 | NP_976050.1 | 4 | 510 | |||
| GenBank mRNA | AK074108.1 | - | 2 | 105 | ||
| BC047949.1 | - | 4 | 510 | |||
| BC137087.1 | - | 4 | 384 | 9021084 | ||
| BC137088.1 | - | 4 | 384 | 9021085 | ||
| BC142952.1 | - | 2 | 316 | |||

FAM102A
Genomic Map