siRNA Details
| Gene Symbol: LOC399815 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC053499.1 | - | 6 | 1023 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC399815| Gene Symbol: LOC399815 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC053499.1 | - | 6 | 1023 | ||