siRNA Details
| Gene Symbol: FOXI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207426.2 | NP_997309.2 | 2 | 734 | ||
| GenBank mRNA | AK128865.1 | - | 2 | 320 | ||
| BC137296.1 | - | 2 | 276 | |||
| BC137297.1 | - | 2 | 276 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FOXI2| Gene Symbol: FOXI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207426.2 | NP_997309.2 | 2 | 734 | ||
| GenBank mRNA | AK128865.1 | - | 2 | 320 | ||
| BC137296.1 | - | 2 | 276 | |||
| BC137297.1 | - | 2 | 276 | |||