siRNA Details
| Gene Symbol: LOC105376554 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135109.1 | 2 | 150 | |||
| GenBank mRNA | AK127718.1 | - | 2 | 150 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105376554| Gene Symbol: LOC105376554 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135109.1 | 2 | 150 | |||
| GenBank mRNA | AK127718.1 | - | 2 | 150 | ||