siRNA Details
| Gene Symbol: PCNX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032223.3 | NP_115599.2 | 2 | 241 | ||
| XM_005273987.4 | XP_005274044.1 | 2 | 2125 | |||
| XM_005273990.4 | XP_005274047.1 | 2 | 2125 | |||
| XM_006718549.3 | XP_006718612.1 | 2 | 2125 | |||
| XM_006718551.3 | XP_006718614.1 | 2 | 2125 | |||
| XM_011545024.2 | XP_011543326.1 | 3 | 1298 | |||
| XM_011545025.2 | XP_011543327.1 | 2 | 2125 | |||
| XM_011545028.2 | XP_011543330.1 | 2 | 2125 | |||
| GenBank mRNA | DY654455.1 | - | 2 | 138 | ||

PCNX3
Genomic Map