siRNA Details
| Gene Symbol: PATE4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144874.1 | NP_001138346.1 | 2 | 118 | ||
| GenBank mRNA | AK123042.1 | - | 2 | 118 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PATE4| Gene Symbol: PATE4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144874.1 | NP_001138346.1 | 2 | 118 | ||
| GenBank mRNA | AK123042.1 | - | 2 | 118 | ||