siRNA Details
| Gene Symbol: LINC00938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028408.1 | 1 | 609 | |||
| GenBank mRNA | BC008245.1 | - | 1 | 186 | ||
| BC037237.1 | - | 1 | 170 | |||
| BC047417.1 | - | 1 | 435 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00938| Gene Symbol: LINC00938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028408.1 | 1 | 609 | |||
| GenBank mRNA | BC008245.1 | - | 1 | 186 | ||
| BC037237.1 | - | 1 | 170 | |||
| BC047417.1 | - | 1 | 435 | |||