siRNA Details
Gene Symbol: LOC100507065 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120431.1 | 2, 3 | 384 | |||
NR_120432.1 | 4, 5 | 557 | ||||
GenBank mRNA | BE895205.1 | - | 342 | |||
CA430002.1 | - | 2, 3 | 475 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100507065 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120431.1 | 2, 3 | 384 | |||
NR_120432.1 | 4, 5 | 557 | ||||
GenBank mRNA | BE895205.1 | - | 342 | |||
CA430002.1 | - | 2, 3 | 475 |