siRNA Details
| Gene Symbol: LOC100507065 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120431.1 | 3 | 521 | |||
| NR_120432.1 | 5 | 694 | ||||
| NR_120433.1 | 5 | 698 | ||||
| GenBank mRNA | BE895205.1 | - | 479 | |||
| BG186681.1 | - | 1 | 385 | |||
| BU619406.1 | - | 1 | 71 | |||
| CA430002.1 | - | 2 | 338 | |||

LOC100507065
Genomic Map