siRNA Details
| Gene Symbol: LINC00173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027345.1 | 2 | 1100 | |||
| GenBank mRNA | AK124947.1 | - | 2 | 1100 | ||
| BC121822.1 | - | 2 | 774 | |||
| BC122872.1 | - | 2 | 790 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00173| Gene Symbol: LINC00173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027345.1 | 2 | 1100 | |||
| GenBank mRNA | AK124947.1 | - | 2 | 1100 | ||
| BC121822.1 | - | 2 | 774 | |||
| BC122872.1 | - | 2 | 790 | |||