siRNA Details
| Gene Symbol: LOC100506016 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134944.1 | 1 | 620 | |||
| GenBank mRNA | BC038751.1 | - | 1 | 621 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506016| Gene Symbol: LOC100506016 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134944.1 | 1 | 620 | |||
| GenBank mRNA | BC038751.1 | - | 1 | 621 | ||