siRNA Details
| Gene Symbol: LOC101927690 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135241.1 | 2 | 287 | |||
| GenBank mRNA | BC037850.1 | - | 2 | 273 | ||
| DA296388.1 | - | 2 | 288 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927690| Gene Symbol: LOC101927690 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135241.1 | 2 | 287 | |||
| GenBank mRNA | BC037850.1 | - | 2 | 273 | ||
| DA296388.1 | - | 2 | 288 | |||