siRNA Details
| Gene Symbol: RNF40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001207033.1 | NP_001193962.1 | 20 | 5545 | ||
| NM_001207034.1 | NP_001193963.1 | 18 | 5248 | |||
| NM_001286572.2 | NP_001273501.1 | 20 | 4909 | |||
| NM_014771.3 | NP_055586.1 | 20 | 5548 | |||

RNF40
Genomic Map