siRNA Details
| Gene Symbol: SYCE1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129979.1 | NP_001123451.1 | 9, 10 | 590 | ||
| GenBank mRNA | BC035731.1 | - | 1, 2 | 104 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SYCE1L| Gene Symbol: SYCE1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129979.1 | NP_001123451.1 | 9, 10 | 590 | ||
| GenBank mRNA | BC035731.1 | - | 1, 2 | 104 | ||