siRNA Details
| Gene Symbol: MED11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001683.3 | NP_001001683.1 | 2 | 244 | ||
| NM_001305000.1 | NP_001291929.1 | 2 | 244 | |||
| GenBank mRNA | AF161414.1 | - | 2 | 219 | ||
| BC070377.1 | - | 2 | 224 | 6450545 | ||
| BG503911.1 | - | 2 | 225 | |||
| HG518163.1 | - | 2 | 239 | |||
| HY036468.1 | - | 2 | 244 | |||

MED11
Genomic Map