siRNA Details
| Gene Symbol: COX10-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_049718.1 | 3 | 1451 | |||
| GenBank mRNA | AK123100.1 | - | 3 | 1451 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

COX10-AS1| Gene Symbol: COX10-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_049718.1 | 3 | 1451 | |||
| GenBank mRNA | AK123100.1 | - | 3 | 1451 | ||