siRNA Details
| Gene Symbol: FLJ35934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104343.1 | 1 | 1478 | |||
| GenBank mRNA | AK093253.1 | - | 1 | 1476 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ35934| Gene Symbol: FLJ35934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104343.1 | 1 | 1478 | |||
| GenBank mRNA | AK093253.1 | - | 1 | 1476 | ||