siRNA Details
| Gene Symbol: LINC01538 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033983.1 | 2 | 240 | |||
| GenBank mRNA | BC042493.1 | - | 2 | 238 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01538| Gene Symbol: LINC01538 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033983.1 | 2 | 240 | |||
| GenBank mRNA | BC042493.1 | - | 2 | 238 | ||