siRNA Details
| Gene Symbol: LINC00683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120419.1 | 2 | 1513 | |||
| GenBank mRNA | AK094957.1 | - | 2 | 421 | ||
| AK127565.1 | - | 2 | 1514 | |||
| Gene Symbol: LOC101927651 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110784.1 | 5 | 863 | |||

LINC00683
Genomic Map