siRNA Details
| Gene Symbol: OVOL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302757.1 | NP_001289686.1 | 2 | 128 | ||
| XM_011527251.2 | XP_011525553.1 | 1 | 22 | |||
| XM_017027190.1 | XP_016882679.1 | 1 | 223 | |||
| XM_017027191.1 | XP_016882680.1 | 2 | 151 | |||

OVOL3
Genomic Map