siRNA Details
| Gene Symbol: ZNF880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145434.1 | NP_001138906.1 | 4 | 507 | ||
| XM_017026812.1 | XP_016882301.1 | 4 | 524 | |||
| GenBank mRNA | AK001357.1 | - | 72 | |||
| AK297834.1 | - | 4 | 473 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF880| Gene Symbol: ZNF880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145434.1 | NP_001138906.1 | 4 | 507 | ||
| XM_017026812.1 | XP_016882301.1 | 4 | 524 | |||
| GenBank mRNA | AK001357.1 | - | 72 | |||
| AK297834.1 | - | 4 | 473 | |||