siRNA Details
| Gene Symbol: ZNF880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145434.1 | NP_001138906.1 | 4 | 648 | ||
| XM_017026812.1 | XP_016882301.1 | 4 | 665 | |||
| GenBank mRNA | AK297834.1 | - | 4 | 614 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF880| Gene Symbol: ZNF880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145434.1 | NP_001138906.1 | 4 | 648 | ||
| XM_017026812.1 | XP_016882301.1 | 4 | 665 | |||
| GenBank mRNA | AK297834.1 | - | 4 | 614 | ||