siRNA Details
| Gene Symbol: ZNF814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144989.1 | NP_001138461.1 | 3 | 2546 | ||
| XM_017027197.1 | XP_016882686.1 | 2 | 2191 | |||
| GenBank mRNA | AK300483.1 | - | 3 | 2512 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF814| Gene Symbol: ZNF814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144989.1 | NP_001138461.1 | 3 | 2546 | ||
| XM_017027197.1 | XP_016882686.1 | 2 | 2191 | |||
| GenBank mRNA | AK300483.1 | - | 3 | 2512 | ||