siRNA Details
| Gene Symbol: ZNF587B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204818.1 | NP_001191747.1 | 3 | 916 | ||
| GenBank mRNA | AK299091.1 | - | 3 | 897 | ||
| U71363.1 | - | 4 | 907 | |||
| Gene Symbol: ZNF814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144989.1 | NP_001138461.1 | 3 | 953 | ||
| XM_017027197.1 | XP_016882686.1 | 2 | 598 | |||

ZNF587B
Genomic Map