siRNA Details
| Gene Symbol: LOC102724312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125994.1 | 2 | 957 | |||
| NR_125995.1 | 2 | 665 | ||||
| NR_125996.1 | 2 | 655 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724312| Gene Symbol: LOC102724312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125994.1 | 2 | 957 | |||
| NR_125995.1 | 2 | 665 | ||||
| NR_125996.1 | 2 | 655 | ||||