siRNA Details
Gene Symbol: SLC35E2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110781.2 | NP_001104251.1 | 9 | 2877 | ||
NM_001290264.1 | NP_001277193.1 | 10 | 3295 | |||
XM_011542072.1 | XP_011540374.1 | 5 | 2238 | |||
GenBank mRNA | AB007916.3 | - | 9 | 2815 | ||
AK092311.1 | - | 3 | 2016 | |||
AK125619.1 | - | 9 | 2801 | |||
BC110653.1 | - | 9 | 2853 | 4392812 | ||
CR936618.1 | - | 10 | 3014 |
Gene Symbol: SLC35E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182838.2 | NP_878258.1 | 6 | 3147 | ||
GenBank mRNA | AK092315.1 | - | 1 | 695 | ||
AK126501.1 | - | 1 | 1074 |
