siRNA Details
Gene Symbol: C1orf141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276351.1 | NP_001263280.1 | 8 | 1307 | ||
NM_001276352.1 | NP_001263281.1 | 9 | 1333 | |||
NR_075077.1 | 10 | 1476 | ||||
XM_011541465.2 | XP_011539767.1 | 9 | 1425 | |||
XM_011541466.2 | XP_011539768.1 | 9 | 1425 | |||
XM_011541467.1 | XP_011539769.1 | 9 | 1395 | |||
XM_011541469.1 | XP_011539771.1 | 5 | 1005 | |||
XM_017001276.1 | XP_016856765.1 | 9 | 1478 |
