siRNA Details
| Gene Symbol: C1orf141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276351.1 | NP_001263280.1 | 8 | 914 | ||
| NM_001276352.1 | NP_001263281.1 | 9 | 940 | |||
| NR_075077.1 | 10 | 1083 | ||||
| XM_011541465.2 | XP_011539767.1 | 9 | 1032 | |||
| XM_011541466.2 | XP_011539768.1 | 9 | 1032 | |||
| XM_011541467.1 | XP_011539769.1 | 9 | 1002 | |||
| XM_011541469.1 | XP_011539771.1 | 5 | 612 | |||
| XM_017001276.1 | XP_016856765.1 | 9 | 1085 | |||
| GenBank mRNA | BC047053.1 | - | 5 | 3263 | ||
| BC090886.1 | - | 7 | 886 | 30378691 | ||

C1orf141
Genomic Map